missing translation for 'onlineSavingsMsg'
Learn More

LY-2584702 tosylate salt, MedChemExpress

Product Code. 16497927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497927 5 mg 5mg
16435208 10 mg 10mg
16425896 100 mg 100mg
16411427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497927 Supplier Medchem Express Supplier No. HY12493A
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress LY-2584702 tosylate salt is a selective ATP competitive inhibitor of p70S6K with an IC50 of 4 nM. In S6K1 enzyme assay, the IC50 of LY-2584702 is 2 nM.

LY-2584702 tosylate salt is a selective ATP competitive inhibitor of p70S6K with an IC50 of 4 nM. In S6K1 enzyme assay, the IC50 of LY-2584702 is 2 nM.

Specifications

Chemical Name or Material LY-2584702 tosylate salt
CAS 1082949-68-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H27F4N7O3S
Quantity 5 mg
Solubility Information DMSO : 10.25 mg/mL (16.60 mM; Need ultrasonic and warming)
SMILES CN1C=C(C2=CC=C(F)C(C(F)(F)F)=C2)N=C1C3CCN(C4=C5C(NN=C5)=NC=N4)CC3.O=S(C6=CC=C(C)C=C6)(O)=O
Molecular Weight (g/mol) 617.62
Formula Weight 617.62
Percent Purity 96.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.