missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ LY 2389575 hydrochloride

Product Code. 18776148
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776148 10 mg 10mg
18756939 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18776148 Supplier R&D Systems™ Supplier No. 4969/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective negative allosteric modulator of mGlu3

LY 2389575 hydrochloride is a selective negative allosteric modulator of mGlu3 (IC50 = 190 nM). Exhibits > 65-fold selectivity for mGlu3 over other mGlu receptors. Abolishes the neuroprotective action of LY 379268 against amyloid β toxicity in mixed cortical neuronal and astrocyte cell cultures.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type LY 2389575 hydrochloride
Molecular Weight (g/mol) 438.58
Product Line Tocris
Inhibitors LY 2389575 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.