missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 2389575 hydrochloride
Description
LY 2389575 hydrochloride is a selective negative allosteric modulator of mGlu3 (IC50 = 190 nM). Exhibits > 65-fold selectivity for mGlu3 over other mGlu receptors. Abolishes the neuroprotective action of LY 379268 against amyloid β toxicity in mixed cortical neuronal and astrocyte cell cultures.
Specifications
Specifications
| Purity | >98% |
| Quantity | 50 mg |
| Product Type | LY 2389575 hydrochloride |
| Molecular Weight (g/mol) | 438.58 |
| Inhibitors | LY 2389575 hydrochloride |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?