missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£160.00 - £300.00
Specifications
| Antigen | Ly-108 |
|---|---|
| Clone | eBio13G3-19D (13G3-19D) |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056048
|
Invitrogen™
M050T03R03-A |
100 μg |
£300.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055480
|
Invitrogen™
M050T02R03-A |
25 μg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly-108 Monoclonal antibody specifically detects Ly-108 in Mouse samples. It is validated for Flow CytometrySpecifications
| Ly-108 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Mouse | |
| Mouse | |
| Q9ET39 | |
| 30925 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBio13G3-19D (13G3-19D) | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
| IgG2a κ | |
| Affinity chromatography | |
| SLAMF6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title