Learn More
Invitrogen™ LXR beta Recombinant Superclonal™ Antibody (8 HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711097
Description
This antibody is predicted to react with Monkey, Cat and Bat Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
Specifications
| LXR beta | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P55055, Q60644 | |
| NR1H2 | |
| Peptides corresponding to Human NR1H2 (aa 244-257, 57-74). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 8 HCLC | |
| Unconjugated | |
| NR1H2 | |
| AI194859; liver X nuclear receptor beta; liver X receptor beta; LX receptor beta; LXR; LXR beta; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; Nr1h2; NR1H2 protein; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; RIP15; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2; UNR; Unr2; UR | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22260, 7376 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.