missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR beta/NR1H2 Antibody (LXRB/2731), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08914DL594
This item is not returnable.
View return policy
Description
LXR beta/NR1H2 Monoclonal antibody specifically detects LXR beta/NR1H2 in Human samples. It is validated for ELISA, Protein Array
Specifications
| LXR beta/NR1H2 | |
| Monoclonal | |
| DyLight 594 | |
| Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta | |
| Recombinant full-length human LXR beta/NR1H2 protein (Uniprot: P55055) | |
| 100 μg | |
| Diabetes Research, Lipid and Metabolism | |
| 7376 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA, Protein Array | |
| LXRB/2731 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction