missing translation for 'onlineSavingsMsg'
Learn More

LXR beta/NR1H2 Antibody (LXRB/2731), DyLight 488, Novus Biologicals™

Product Code. 18580075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18580075 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18580075 Supplier Novus Biologicals Supplier No. NBP308914G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LXR beta/NR1H2 Monoclonal antibody specifically detects LXR beta/NR1H2 in Human samples. It is validated for ELISA, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR beta/NR1H2
Applications ELISA, Protein Array
Classification Monoclonal
Clone LXRB/2731
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta
Host Species Mouse
Immunogen Recombinant full-length human LXR beta/NR1H2 protein (Uniprot: P55055)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7376
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.