missing translation for 'onlineSavingsMsg'
Learn More

LXR beta/NR1H2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18404767 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404767 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404767 Supplier Novus Biologicals Supplier No. NBP176256AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LXR beta/NR1H2 Polyclonal antibody specifically detects LXR beta/NR1H2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR beta/NR1H2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus human LXR-B. The immunogen is located within the first 50 amino acids of LXR-B. Amino Acid Squence: SPTVKEEGPEPWPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7376
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.