missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Antibody (OTI1A5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72550C
This item is not returnable.
View return policy
Description
LXR alpha/NR1H3 Monoclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LXR alpha/NR1H3 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NR1H3 | |
| Recombinant protein fragment corresponding to amino acids 69-329 of human LXR alpha/NR1H3 (NP_005684) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1A5 | |
| Western Blot, Immunohistochemistry | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| Mouse | |
| Affinity Purified | |
| Diabetes Research, Lipid and Metabolism | |
| 10062.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction