missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LXR alpha Monoclonal Antibody (PPZ0412)
Mouse Monoclonal Antibody
Brand: Invitrogen™ 418300
This item is not returnable.
View return policy
Description
This antibody specifically recognizes human LXR alpha and cross reacts with mouse and rat LXR alpha. This antibody does not recognize human LXR beta.
NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiquitination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.
Specifications
| LXR alpha | |
| Monoclonal | |
| 1 mg/mL | |
| Physiological saline with 0.1% sodium azide | |
| Q13133, Q62685, Q9Z0Y9 | |
| NR1H3 | |
| Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with E.coli-expressed recombinant human LXR-alpha (164-447 aa). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| ChIP Assay, ELISA, Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| PPZ0412 | |
| Unconjugated | |
| NR1H3 | |
| AU018371; liver X nuclear receptor alpha variant 1; liver X receptor alpha; LXR; LXR alpha; Lxra; LXR-a; LXRalpha; LXRb; NER; NERI; NR1H2; Nr1h3; nuclear orphan receptor LXR-alpha; nuclear oxysterol receptor LxR-alpha; nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; oxysterols receptor LXR-alpha; RLD1; RLD-1; ubiquitously-expressed nuclear receptor 1; Unr1 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10062, 22259, 58852 | |
| Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction