missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/beta Polyclonal Antibody

LXR alpha/beta Polyclonal Antibody
Brand: Invitrogen PA19066
This item is not returnable.
View return policy
Description
LXR alpha/beta Polyclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
| LXR alpha/beta | |
| Polyclonal | |
| Unconjugated | |
| NR1H3 | |
| AI194859; AU018371; liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X Receptor; liver X receptor alpha; Liver X receptor beta; LX receptor beta; LXR; LXR alpha; LXR beta; LXRA; LXR-a; LXRalpha; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; NR1H2; NR1H3; nuclear orphan receptor LXR-alpha; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; retinoid X receptor-interacting protein No.15; Rip15; RLD1; RLD-1; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 1; ubiquitously-expressed nuclear receptor 2; Unr; Unr1; Unr2; UR | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10062, 7376 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55055, Q13133 | |
| NR1H2, NR1H3 | |
| Internal sequence corresponding to amino acids CRLQDKKLPPLLSEI. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction