missing translation for 'onlineSavingsMsg'
Learn More

LX7101, MedChemExpress

Product Code. 16495278
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495278 10 mg 10mg
16461487 50 mg 50mg
16457997 5 mg 5mg
16430176 2 mg 2mg
16425946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495278 Supplier Medchem Express Supplier No. HY12659
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LX7101 is a potent inhibitor of LIMK and ROCK2 with IC50 values of 24, 1.6 and 10 nM for LIMK1, LIMK2 and ROCK2, respectively; also inhibits PKA with an IC50 less than 1 nM.

LX7101 is a potent inhibitor of LIMK and ROCK2 with IC50 values of 24, 1.6 and 10 nM for LIMK1, LIMK2 and ROCK2, respectively

Specifications

Chemical Name or Material LX7101
CAS 1192189-69-7
Color White
Purity Grade Notes Research
Molecular Formula C23H29N7O3
Quantity 10 mg
Solubility Information DMSO : 150 mg/mL (332.21 mM; Need ultrasonic)
SMILES NCC1(C(NC2=CC(OC(N(C)C)=O)=CC=C2)=O)CCN(C3=C4C(NC=C4C)=NC=N3)CC1
Molecular Weight (g/mol) 451.52
Formula Weight 451.52
Percent Purity 98.12%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.