missing translation for 'onlineSavingsMsg'
Learn More

LX2343, MedChemExpress

Product Code. 16464438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464438 10 mg 10mg
16455326 100 mg 100mg
16410757 50 mg 50mg
16407167 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464438 Supplier Medchem Express Supplier No. HY111383
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance.

LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance.

Specifications

Chemical Name or Material LX2343
CAS 333745-53-2
Color White
Purity Grade Notes Research
Molecular Formula C22H19ClN2O6S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (210.57 mM)
SMILES O=C(NC1=CC=C(OCO2)C2=C1)CN(C3=CC(Cl)=CC=C3OC)S(=O)(C4=CC=CC=C4)=O
Molecular Weight (g/mol) 474.91
Formula Weight 474.91
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.