missing translation for 'onlineSavingsMsg'
Learn More

CTS™ CTS™ LV-MAX™ Transfection Kit

Product Code. 17143189
Change view
Click to view available options
Quantity:
1 Kit (For 1 L of LV Production)
1 Kit (For 10 L of LV Production)
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17143189 1 Kit (For 1 L of LV Production) 1 L
17153189 1 Kit (For 10 L of LV Production) 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17143189 Supplier CTS™ Supplier No. A4132601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Gibco CTS LV-MAX Transfection Kit is an essential component of the CTS LV-MAX lentiviral production system.

The Gibco CTS LV-MAX Transfection Kit is an essential component of the CTS LV-MAX lentiviral production system. Based on extensive Design of Experiment (DOE) methodologies, the cationic lipid-based transfection reagent, lentiviral production supplement, and LV enhancer are specifically screened for maximal delivery of multiple DNA plasmids to achieve the highest possible lentiviral titer when used in conjunction with CTS Viral Production Cells cultured in CTS LV-MAX Production Medium.

All CTS LV-MAX Transfection Kit components are produced under cGMP manufacturing conditions with extensive quality-control testing to ensure proper preclinical, clinical, and commercial therapeutic viral vector applications.

CTS LV-MAX Transfection Kit features include:

  • Uniquely designed to enhance transient transfection of multiple DNA plasmids into high density suspension HEK293-derived cell culture
  • Enables high viral titers of up to 1 x 108 TU/mL (unconcentrated LVV-GFP) to allow for a cost-effective alternative to PEI-based transfection reagents
  • Scalable protocol for 96-deep well blocks, flasks, and bioreactors
  • Regulatory-compliant reagents produced in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.

The CTS LV-MAX Transfection Kit requires CTS Opti-MEM I (not included in this kit) for complexation. The chemically-defined, serum-free, animal origin-free features of the CTS LV-MAX Transfection Kit result in high reproducibility and lot-to-lot reliability. The formulation of this product is identical to the research-grade LV-MAX Transfection Kit, but the CTS LV-MAX Transfection Kit is validated for clinical and commercial therapeutic applications.

Custom configuration and packaging available upon request.

TRUSTED_SUSTAINABILITY

Tekniske data

Classification Animal Origin-Free
Content And Storage • CTS LV-MAX Supplement, 50 mL
• CTS LV-MAX Transfection Reagent, 6 mL
• CTS LV-MAX Enhancer, 40 mL

Store at 2–8°C and protect from light. Ships on gel ice.

Culture Type Mammalian
Cell Type HEK293
Format Bottle
For Use With (Application) LV Production
Product Type Transfection Kit
Quantity 1 Kit (For 1 L of LV Production)
Reagent Type Cationic Lipid-Based Transfection
Shipping Condition Gel Ice
Manufacturing Quality ISO 13485, FDA-registered, 21 CFR 820
Vis mere Vis mindre

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.