missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LV-MAX™ Lentiviral Packaging Mix

Product Code. 31784130 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31784130 1.5 mL 1.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31784130 Supplier Invitrogen™ Supplier No. A43237
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LV-MAX Lentiviral Packaging Mix is an optimized single formulation containing a defined mixture of three packaging plasmids: pLP1, pLP2, and pLP/VSVG.

The LV-MAX Lentiviral Packaging Mix is a single, optimized set of packaging plasmids, designed to streamline and enhance the production of lentiviral vectors. This mix contains a defined combination of three essential packaging plasmids: pLP1, pLP2, and pLP/VSVG, which together supply the necessary structural and replication proteins in trans when expressed in HEK293-derived Viral Production Cells. This third-generation packaging system offers enhanced biosafety, as the Rev gene is supplied on a separate plasmid, independent of other lentiviral genes, thereby significantly reducing the likelihood of recombination and producing replication-competent lentiviral particles.

Key features of the LV-MAX Lentiviral Packaging Mix

  • Optimized formulation—combines three packaging plasmids (pLP1, pLP2, and pLP/VSVG) into a single mix, saving process development time by minimizing the need to optimize plasmid ratios
  • High titer—achieves high titers of lentiviral vectors when used with the LV-MAX Lentiviral Production System

Applications

  • Gene therapy—ideal for the production of lentiviral vectors for gene therapy research and development
  • Cell therapy—supports the generation of lentiviral vectors for cell therapy applications, facilitating seamless transition from bench to clinic
  • Research and development—provides a reliable and scalable solution for viral vector production in various research settings
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lentiviral Packaging Mix
Sterility Non-sterile
Quantity 1.5 mL
Content And Storage Store at -5 to -30°C. Ships on gel ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.