missing translation for 'onlineSavingsMsg'
Learn More

LUZP4 Antibody (OTI6D11), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18430107 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430107 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430107 Supplier Novus Biologicals Supplier No. NBP272549AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LUZP4 Monoclonal antibody specifically detects LUZP4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUZP4
Applications Western Blot
Classification Monoclonal
Clone OTI6D11
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias leucine zipper protein 4
Host Species Mouse
Immunogen Full length recombinant protein of human LUZP4 (NP_057467) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51213
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.