missing translation for 'onlineSavingsMsg'
Learn More

Luzindole, TRC

Product Code. 30483519
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483519 5 mg 5mg
30473370 10 mg 10mg
30471342 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483519 Supplier Toronto Research Chemicals Supplier No. TRCL4820005MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117946-91-5
Molecular Formula C19 H20 N2 O
Molecular Weight (g/mol) 292.37
IUPAC Name N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCc1c(Cc2ccccc2)[nH]c3ccccc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H20N2O/c1-14(22)20-12-11-17-16-9-5-6-10-18(16)21-19(17)13-15-7-3-2-4-8-15/h2-10,21H,11-13H2,1H3,(H,20,22)
Formula Weight 292.1576
Chemical Name or Material Luzindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.