missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Luzindole

Product Code. 18064622
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064622 10 mg 10mg
18084072 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064622 Supplier R&D Systems™ Supplier No. 0877/10
  • ONLINE EXCLUSIVE PRICE
    £193.30
    £173.25 / 10mg
    Save £20.05
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Competitive melatonin MT1/MT2 antagonist

Melatonin antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117946-91-5
Molecular Formula C19H20N2O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 292.38
Purity >99%
Molecular Weight (g/mol) 292.38
Chemical Name or Material Luzindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.