missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Luxol™ Fast Blue MBSN, pure

Product Code. 10591374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591374 100 g Glass bottle 100g
10348250 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10591374 Supplier Thermo Scientific Chemicals Supplier No. 212171000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1328-51-4
Molecular Formula C32H14CuN8Na2O6S2
Molecular Weight (g/mol) 780.16
MDL Number MFCD00071424
InChI Key DKBXPLYSDKSFEQ-UHFFFAOYSA-L
Synonym Solvent Blue 38
PubChem CID 92030797
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]

Specifications

Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acohols like methanol, ethanol, propano, ls; very soluble in gylcols, ethanolamine, choline, . insoluble in hydrocarbons.
Infrared Spectrum Authentic
Formula Weight 778.15
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Blue
Quantity 100 g
Physical Form Powder
Chemical Name or Material Luxol™ Fast Blue MBSN, Pure

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.