Learn More
Invitrogen™ Lutikizumab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560126
 View more versions of this product
Product Code. 30284092
  • £2005.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lutikizumab is a biosimilar that targets IL1A .
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lutikizumab Humanized
Recombinant Monoclonal
Unconjugated
ABT-981
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized