missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11392508
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11392508

Brand: Thermo Scientific Alfa Aesar 014566.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lutetium Sulfate also called Lutetium Sulphate, is applied in making phosphors. Stable Lutetium can be used as catalysts in petroleum cracking in refineries and can also be used in alkylation, hydrogenation, and polymerization applications. It can also be used as an ideal host for X-ray phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13473-77-3
MDL Number MFCD00011499

Specifications

Physical Form Crystalline
Quantity 1 g
Merck Index 14,5615
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 782.24 (683.12 Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) sulfate octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.