missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11366318
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
12032-20-1
Lu2O3
397.93
MFCD00011100
UGBIHFMRUDAMBY-UHFFFAOYSA-N
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
dilutetium(3+) trioxidandiide
[O--].[O--].[O--].[Lu+3].[Lu+3]
This item is not returnable. View return policy
12032-20-1
Lu2O3
397.93
MFCD00011100
UGBIHFMRUDAMBY-UHFFFAOYSA-N
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
dilutetium(3+) trioxidandiide
[O--].[O--].[O--].[Lu+3].[Lu+3]

Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2487°C
Physical Form Powder
Quantity 25 g
Merck Index 14,5615
Solubility Information Insoluble in water.
Formula Weight 397.94
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 9.420
Chemical Name or Material Lutetium(III) oxide, REacton™, (REO)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'