missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) oxide, 99.9% (REO), Thermo Scientific™

Product Code. 11393298
Change view
Click to view available options
Quantity:
2 g
5 g
Unit Size:
2g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393298 5 g 5g
11303308 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393298 Supplier Thermo Scientific Alfa Aesar Supplier No. 036244.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Melting Point 2487°C
Physical Form Sintered Lump
Quantity 5 g
Merck Index 14,5615
Solubility Information Insoluble in water.
Formula Weight 397.94
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 9.420 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.