missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11420470
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100641-16-5
LuN3O9
360.98
MFCD00150421
APRNQTOXCXOSHO-UHFFFAOYSA-N
lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
16212852
[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100641-16-5
LuN3O9
360.98
MFCD00150421
APRNQTOXCXOSHO-UHFFFAOYSA-N
lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
16212852
[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Lutetium(III) nitrate hydrate is used in ceramics, glass, phosphors and in lasers. It finds application as a catalyst in petroleum cracking and to prepare laser crystal. Further, it is used in hydrogenation, alkylation and polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula Lu(NO3)3·xH2O
UN Number UN1477
Solubility Information Soluble in water. Slightly soluble in strong mineral acids.
IUPAC Name lutetium(3+);trinitrate;hydrate
Formula Weight 360.98 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'