missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lutetium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Specifications
Melting Point | 1050°C |
Physical Form | Flakes or Powder |
Quantity | 5 g |
Solubility Information | Soluble in chloroform,carbon tetrachloride and carbon disulfide. |
Formula Weight | 555.68 |
Percent Purity | 99.9% |
Packaging | Ampule |
Sensitivity | Hygroscopic |
Density | 5.60 g/mL |
Assay | (REO) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'