missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals

£138.00 - £456.00

Chemical Identifiers

CAS 10099-66-8
MDL Number MFCD00011099
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378697
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035802.03
1 g
£138.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11388697
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035802.06
5 g
£456.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as an additive for chemical industry, single crystal scintillators, carbon arc rods, PET scan detectors, high refractive index glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10099-66-8
MFCD00011099

Specifications

905°C (sublimation)
Powder
14,5615
281.33
Ampule
3.98 g/mL
Lutetium(III) chloride, Ultra dry
10099-66-8
MFCD00011099
Soluble in water.
99.98%
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

missing translation for 'einecsNumber' : 233-240-1

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only