missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific™

Product Code. 10167882 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167882 5 g Glass bottle 5g
10786373 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167882 Supplier Thermo Scientific Acros Supplier No. 383540050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-66-8
Molecular Formula Cl3Lu
MDL Number MFCD00011099

Specifications

Color White to Gray
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9%
Linear Formula LuCl3
Formula Weight 281.33
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lutetium(III) chloride
compliance-icons
Product Identifier
  • Lutetium(III) chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.