missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Product Code. 11332288
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332288 5 g 5g
11322288 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332288 Supplier Thermo Scientific Alfa Aesar Supplier No. 013189.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17966-84-6 | C15H21LuO6 | 472.29 g/mol

Lutetium(III) 2,4-pentanedionate is involved in refluxing 1,2,4-trichlorobenzene. It is also used as catalyst of organic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17966-84-6
Molecular Formula C15H21LuO6
Molecular Weight (g/mol) 472.29
MDL Number MFCD00078030
InChI Key HXFHOORFYPCGNF-UHFFFAOYSA-N
Synonym lutetium iii acetylacetonate hydrate, lutetium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy lutetio oxy pent-3-en-2-one
PubChem CID 24282830
SMILES [Lu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;lutetium
Formula Weight 472.3
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.