missing translation for 'onlineSavingsMsg'
Learn More

Lutetium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11331288
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7439-94-3
Lu
174.97
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
23929
CHEBI:33382
lutetium
[Lu]
7439-94-3
Lu
174.97
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
23929
CHEBI:33382
lutetium
[Lu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Silver to Gray
UN Number UN3178
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.62g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Lutetium foil, 0.1mm (0.004 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'