missing translation for 'onlineSavingsMsg'
Learn More

Lutetium, AAS standard solution, Specpure™ Lu 1000μg/mL

Product Code. 11463840
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463840 100 mL 100mL
11396399 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463840 Supplier Thermo Scientific Alfa Aesar Supplier No. 089886.AE
  • ONLINE EXCLUSIVE PRICE
    £55.10
    £49.65 / 100mL
    Save £5.45
    9% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1 , 7697-37-2 , 7732-18-5
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Type Analytical Standard
Quantity 100 mL
Grade AAS
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Lutetium(III) oxid: 0.11%; Water: 94.89%
Chemical Name or Material Lutetium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.