missing translation for 'onlineSavingsMsg'
Learn More

Luteolin, TRC

Product Code. 30007521
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007521 100 mg 100mg
30008738 10 mg 10mg
30005703 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007521 Supplier Toronto Research Chemicals Supplier No. TRCL475000100MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-70-3
Molecular Formula C15 H10 O6
Molecular Weight (g/mol) 286.24
Synonym 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Flavone, 3',4',5,7-tetrahydroxy- (8CI), Luteolin (6CI), 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-benzopyran-4-one, 3',4',5,7-Tetrahydroxyflavone, 5,7,3',4'-Tetrahydroxyflavone, Cyanidenon, Cyanidenon 1470, Digitoflavone, Flacitran, Luteolin-7-O-glucopyranosyl 8-C-(6' '-acetyl)-glucopyranoside, Luteoline, Luteolol, Salifazide, Weld lake, Yama Kariyasu
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
SMILES Oc1cc(O)c2C(=O)C=C(Oc2c1)c3ccc(O)c(O)c3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H10O6/c16-8-4-11(19)15-12(20)6-13(21-14(15)5-8)7-1-2-9(17)10(18)3-7/h1-6,16-19H
Formula Weight 286.0477
Chemical Name or Material Luteolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.