missing translation for 'onlineSavingsMsg'
Learn More

Luteolin, MedChemExpress

Product Code. 16414777
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16414777

Brand: Medchem Express HYN0162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Luteolin (Luteoline), a flavanoid compound, is a potent Nrf2 inhibitor. Luteolin has anti-inflammatory, anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, including human non-small lung cancer cells.

Luteolin (Luteoline), a flavanoid compound, is a potent Nrf2 inhibitor. Luteolin has anti-inflammatory, anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, including human non-small lung cancer cells.

Specifications

Chemical Name or Material Luteolin
CAS 491-70-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O6
Quantity 50 mg
Synonym Luteoline Luteolol Digitoflavone
Solubility Information DMSO : ≥ 100 mg/mL (349.36 mM)
SMILES O=C1C=C(C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 286.24
Formula Weight 286.24
Percent Purity 96.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.