missing translation for 'onlineSavingsMsg'
Learn More

Luteolin, MedChemExpress

Product Code. 16472356
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472356 500 mg 500mg
16414777 50 mg 50mg
16409016 100 mg 100mg
16408888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472356 Supplier Medchem Express Supplier No. HYN0162
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Luteolin (Luteoline), a flavanoid compound, is a potent Nrf2 inhibitor. Luteolin has anti-inflammatory, anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, including human non-small lung cancer cells.

Luteolin (Luteoline), a flavanoid compound, is a potent Nrf2 inhibitor. Luteolin has anti-inflammatory, anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, including human non-small lung cancer cells.

Specifications

Chemical Name or Material Luteolin
CAS 491-70-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O6
Quantity 500 mg
Synonym Luteoline Luteolol Digitoflavone
Solubility Information DMSO : ≥ 100 mg/mL (349.36 mM)
SMILES O=C1C=C(C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 286.24
Formula Weight 286.24
Percent Purity 96.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.