missing translation for 'onlineSavingsMsg'
Learn More

Luteolin 7-O-glucuronide, MedChemExpress

Product Code. 16481638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481638 5 mg 5mg
16449228 10 mg 10mg
16423926 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481638 Supplier Medchem Express Supplier No. HYN1463
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Luteolin 7-O-glucuronide could inhibit Matrix Metalloproteinases (MMP) activities, with IC50s of 17.63, 7.99, 11.42, 12.85, 0.03 μM for MMP-1, MMP-3, MMP-8, MMP-9, MMP-13, respectively.

Luteolin 7-O-glucuronide could inhibit Matrix Metalloproteinases (MMP) activities, with IC50s of 17.63, 7.99, 11.42, 12.85, 0.03 μM for MMP-1, MMP-3, MMP-8, MMP-9, MMP-13, respectively.

Specifications

Chemical Name or Material Luteolin 7-O-glucuronide
CAS 29741-10-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H18O12
Quantity 5 mg
Synonym Luteolin 7-glucuronide
Solubility Information DMSO : 125 mg/mL (270.35 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](C(O)=O)O1)O)O)[C@@H]1OC2=CC(O)=C3C(C=C(C4=CC=C(O)C(O)=C4)OC3=C2)=O
Molecular Weight (g/mol) 462.36
Formula Weight 462.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.