missing translation for 'onlineSavingsMsg'
Learn More

Luteolin-3-O-beta-D-glucuronide, MedChemExpress

Product Code. 16441738
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441738 5 mg 5mg
16409308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441738 Supplier Medchem Express Supplier No. HYN4099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Luteolin-3-O-beta-D-glucuronide is a luteolin glucosiduronic acid consisting of luteolin having a beta-D-glucosiduronic acid residue attached at the 3'-position.

Luteolin-3-O-beta-D-glucuronide is a luteolin glucosiduronic acid consisting of luteolin having a beta-D-glucosiduronic acid residue attached at the 3'-position.

Specifications

Chemical Name or Material Luteolin-3-O-beta-D-glucuronide
CAS 53527-42-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H18O12
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (216.28 mM; Need ultrasonic)
SMILES O=C1C2=C(O)C=C(O)C=C2OC(C3=CC(O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(O)=O)=C(O)C=C3)=C1
Molecular Weight (g/mol) 462.36
Formula Weight 462.36
Percent Purity 98.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.