missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Luteinizing Hormone Releasing Hormone (LH-RH), salmon

Product Code. 16462709
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462709 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462709 Supplier MedChemExpress Supplier No. HYP0243
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Luteinizing Hormone Releasing Hormone (LH-RH), salmon (Salmon GnRH) is the hypophysiotropic decapeptide synthesized in the hypothalamus that plays a crucial role in the control of reproductive functions.

Specifications

Purity 98.07%
CAS 86073-88-3
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1212.31 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature
Synonym Salmon GnRH∣ Salmon gonadotropin-releasing hormone∣ sGnRH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.