missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Luteinizing Hormone-beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109907
This item is not returnable.
View return policy
Description
Immunogen sequence: MRVLQAVLPP LPQVVCTYRD VRFESIRLPG CPRGVDPVVS FPVALSCRCG PCRRSTSDCG GPKDHPLTCD HPQLSGLLFL.
This gene is a member of the glycoprotein hormone beta chain family and encodes the beta subunit of luteinizing hormone (LH). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
Specifications
| Luteinizing Hormone-beta | |
| Polyclonal | |
| Unconjugated | |
| LHB | |
| aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; CGA; Cga1; CG-alpha; CGB4; Choriogonadotropin alpha chain; Chorionic gonadotrophin subunit alpha; Follicle-stimulating hormone alpha chain; follitropin; Follitropin alpha chain; FSHA; FSH-alpha; FSH-B; FSH-beta; Glycoprotein hormones alpha chain; GPHa; GPHA1; GPHalpha; HCG; HH23; hLHB; Interstitial cell-stimulating hormone; LH; LH alpha 1-1; LH alpha 1-2; LH alpha 1-3; LH alpha 3; LH beta-1; LH beta-2; LH beta-3; LHA; Lhb; LH-B; LSH-alpha; LSH-B; LSH-beta; Luteinizing hormone alpha chain; Luteinizing hormone subunit beta; Lutropin alpha chain; Lutropin beta chain; Lutropin subunit beta; Thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; TSHA; TSH-alpha | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3972 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P01229 | |
| LHB | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction