missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luteinizing Hormone Monoclonal Antibody (5304)
GREENER_CHOICE

Product Code. 12807952
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807952 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12807952 Supplier Invitrogen™ Supplier No. MBL0110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MBL0110 targets Luteinizing Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MBL0110 immunogen is human Luteinizing Hormone. MBL0110 detects Luteinizing Hormone which has a predicted molecular weight of approximately 13 kDa. MBL0110 was formerly sold as a Seradyn product.

LH is a member of the glycoprotein hormone family, Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Luteinizing Hormone
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 5304
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LHB
Gene Accession No. P01215, P01229
Gene Alias aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; CGA; Cga1; CG-alpha; CGB4; Choriogonadotropin alpha chain; chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin; follitropin alpha chain; FSHA; FSH-alpha; FSH-B; FSH-beta; glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; HH23; hLHB; interstitial cell stimulating hormone, beta chain; interstitial cell-stimulating hormone; leutenising hormone beta; leutropin; LH; LH alpha 1-1; LH alpha 1-2; LH alpha 1-3; LH alpha 3; LH beta-1; LH beta-2; LH beta-3; LHA; LHB; LH-B; LHbeta; LSH-alpha; LSH-B; LSH-beta; Luteinizing hormone (lutropin) subunit beta; luteinizing hormone alpha chain; luteinizing hormone beta; luteinizing hormone beta polypeptide; luteinizing hormone beta subunit; luteinizing hormone subunit beta; Lutropin alpha chain; lutropin beta chain; Lutropin subunit beta; pituitary hormone alpha subunit; precursor polypeptide; precursor polypeptide (AA -24 to 96); thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; TSHA; TSH-alpha
Gene Symbols Cga, LHB
Host Species Mouse
Immunogen Human Luteinizing Hormone.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081, 3972
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.