missing translation for 'onlineSavingsMsg'
Learn More

Lusutrombopag, MedChemExpress

Product Code. p-200100805
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480326

Brand: Medchem Express HY19883

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Lusutrombopag is an orally bioavailable thrombopoietin (TPO) receptor agonist, used for treatment of chronic liver disease.

Lusutrombopag is an orally bioavailable thrombopoietin (TPO) receptor agonist, used for treatment of chronic liver disease.

Specifications

Chemical Name or Material Lusutrombopag
CAS 1110766-97-6
Color White
Purity Grade Notes Research
Molecular Formula C29H32Cl2N2O5S
Quantity 2 mg
Synonym S-888711
Solubility Information DMSO : ≥ 33 mg/mL (55.79 mM)
SMILES O=C(O)/C(C)=C/C1=C(Cl)C=C(C(NC2=NC(C3=CC=CC([C@@H](OCCCCCC)C)=C3OC)=CS2)=O)C=C1Cl
Molecular Weight (g/mol) 591.55
Formula Weight 591.55
Percent Purity 98.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.