missing translation for 'onlineSavingsMsg'
Learn More

Lusutrombopag, MedChemExpress

Product Code. 16480326
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480326 2 mg 2mg
16467948 10 mg 10mg
16453706 25 mg 25mg
16447746 100 mg 100mg
16433887 50 mg 50mg
16400418 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16480326 Supplier Medchem Express Supplier No. HY19883

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Lusutrombopag is an orally bioavailable thrombopoietin (TPO) receptor agonist, used for treatment of chronic liver disease.

Lusutrombopag is an orally bioavailable thrombopoietin (TPO) receptor agonist, used for treatment of chronic liver disease.

Specifications

Chemical Name or Material Lusutrombopag
CAS 1110766-97-6
Color White
Purity Grade Notes Research
Molecular Formula C29H32Cl2N2O5S
Quantity 2 mg
Synonym S-888711
Solubility Information DMSO : ≥ 33 mg/mL (55.79 mM)
SMILES O=C(O)/C(C)=C/C1=C(Cl)C=C(C(NC2=NC(C3=CC=CC([C@@H](OCCCCCC)C)=C3OC)=CS2)=O)C=C1Cl
Molecular Weight (g/mol) 591.55
Formula Weight 591.55
Percent Purity 98.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.