missing translation for 'onlineSavingsMsg'
Learn More

Melford Luria Broth Millers Modified

Product Code. p-200087005
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018426

Brand: Melford L24080500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For propagation and maintenace of E.

For propagation and maintenace of E.Coli. Low Salt Formula. Molecular Biology Grade.

Dissolve 15.5 grams in 1 liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Peptone: 10 g/L

Yeast Extract: 5 g/L

Sodium Chloride: 0.5 g/L

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.