missing translation for 'onlineSavingsMsg'
Learn More

Lurbinectedin, MedChemExpress

Product Code. 16460276
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
2 mg
Unit Size:
1mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460276 2 mg 2mg
16411609 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460276 Supplier Medchem Express Supplier No. HY16293
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lurbinectedin (PM01183) is a DNA minor groove covalent binder with potent anti-tumour activity; inhibits RMG1 and RMG2 cell growth with IC50 values of 1.25 and 1.16 nM, respectively.

Lurbinectedin (PM01183) is a DNA minor groove covalent binder with potent anti-tumour activity

Specifications

Chemical Name or Material Lurbinectedin
CAS 497871-47-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C41H44N4O10S
Quantity 2 mg
Synonym PM01183
Solubility Information DMSO : 20 mg/mL (25.48 mM; Need ultrasonic)
SMILES O[C@@H]([C@@](CC1=CC(C)=C2OC)([H])N(C)[C@]3([H])C1=C2O)N([C@](COC4=O)([H])C5=C(OCO6)C6=C7C)[C@@]3([H])[C@@](SC[C@@]4(NCC8)C9=C8C%10=CC(OC)=CC=C%10N9)([H])C5=C7OC(C)=O
Molecular Weight (g/mol) 784.87
Formula Weight 784.87
Percent Purity 97.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.