missing translation for 'onlineSavingsMsg'
Learn More

Lurasidone metabolite 14326 hydrochloride, MedChemExpress

Product Code. p-200101244
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16412449

Brand: Medchem Express HYG0002A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lurasidone metabolite 14326 (hydrochloride) is an active metabolite of the atypical antipsychotic Lurasidone.

Lurasidone metabolite 14326 (hydrochloride) is an active metabolite of the atypical antipsychotic Lurasidone.

Specifications

Chemical Name or Material Lurasidone metabolite 14326 hydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C28H37ClN4O3S
Quantity 1 mg
SMILES O=C([C@]([C@@H]1C[C@H](O)[C@H]2C1)([H])[C@]2([H])C3=O)N3C[C@H]4[C@H](CN(CC5)CCN5C6=NSC7=C6C=CC=C7)CCCC4.Cl
Molecular Weight (g/mol) 545.14
Formula Weight 545.14
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.