missing translation for 'onlineSavingsMsg'
Learn More

Lurasidone metabolite 14326 hydrochloride, MedChemExpress

Product Code. 16412449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412449 1 mg 1mg
16411218 5 mg 5mg
16408788 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16412449 Supplier Medchem Express Supplier No. HYG0002A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lurasidone metabolite 14326 (hydrochloride) is an active metabolite of the atypical antipsychotic Lurasidone.

Lurasidone metabolite 14326 (hydrochloride) is an active metabolite of the atypical antipsychotic Lurasidone.

Specifications

Chemical Name or Material Lurasidone metabolite 14326 hydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C28H37ClN4O3S
Quantity 1 mg
SMILES O=C([C@]([C@@H]1C[C@H](O)[C@H]2C1)([H])[C@]2([H])C3=O)N3C[C@H]4[C@H](CN(CC5)CCN5C6=NSC7=C6C=CC=C7)CCCC4.Cl
Molecular Weight (g/mol) 545.14
Formula Weight 545.14
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.