missing translation for 'onlineSavingsMsg'
Learn More

Lupeol, MedChemExpress

Product Code. 16499136
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499136 100 mg 100mg
16443906 25 mg 25mg
16409108 10 mg 10mg
16404897 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499136 Supplier Medchem Express Supplier No. HYN0790
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lupeol (Clerodol; Monogynol B; Fagarasterol) is an active pentacyclic triterpenoid, has anti-oxidant, anti-mutagenic, anti-tumor and anti-inflammatory activity. Lupeol is a potent androgen receptor (AR) inhibitor and can be used for cancer research, especially prostate cancer of androgen-dependent phenotype (ADPC) and castration resistant phenotype (CRPC).

Lupeol (Clerodol

Specifications

Chemical Name or Material Lupeol
CAS 545-47-1
Color White
Purity Grade Notes Research
Molecular Formula C30H50O
Quantity 100 mg
Synonym Clerodol Monogynol B Fagarasterol
Solubility Information Ethanol : 14.29 mg/mL (33.49 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 2 mg/mL (4.69 mM; Need ultrasonic)
SMILES C[C@]1([C@@]2(CC[C@@]3([H])C4(C)C)C)[C@@](CC[C@]2([H])[C@]3(CC[C@@H]4O)C)([H])[C@]([C@@H]5C(C)=C)([H])[C@](C)(CC5)CC1
Molecular Weight (g/mol) 426.72
Formula Weight 426.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.