missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lupeol, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Description
Lupeol (Clerodol
Specifications
Specifications
| Chemical Name or Material | Lupeol |
| CAS | 545-47-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H50O |
| Quantity | 10 mg |
| Synonym | Clerodol Monogynol B Fagarasterol |
| Solubility Information | Ethanol : 14.29 mg/mL (33.49 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 2 mg/mL (4.69 mM; Need ultrasonic) |
| SMILES | C[C@]1([C@@]2(CC[C@@]3([H])C4(C)C)C)[C@@](CC[C@]2([H])[C@]3(CC[C@@H]4O)C)([H])[C@]([C@@H]5C(C)=C)([H])[C@](C)(CC5)CC1 |
| Molecular Weight (g/mol) | 426.72 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction