missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lunsekimig Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554932
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA Isotype of this product is: (sc-5-VH).
Lunsekimig acts by inhibiting the enzyme responsible for the synthesis of a specific cellular component, thereby disrupting the normal function and replication of target cells.
Specifications
| Lunsekimig Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NC30TSLPALB; PRO0903; PRO1708; PRO2044; PRO2619; PRO2675 | |
| Recombinant Human IL13 and TSLP and ALB. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1.7 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| VH-VH |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction