missing translation for 'onlineSavingsMsg'
Learn More

Lung Cancer Marker Antibody (MOC-52) - Azide and BSA Free, Novus Biologicals™

Product Code. 18397906 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397906 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397906 Supplier Novus Biologicals Supplier No. NBP311523

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lung Cancer Marker MOC-52 Monoclonal antibody specifically detects Lung Cancer Marker MOC-52 in Human samples. It is validated for Immunofluorescence, Immunofluorescence, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lung Cancer Marker MOC-52
Applications Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone MOC-52
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml, Immunocytochemistry 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Host Species Mouse
Immunogen Lung Cancer-associated antigen isolated from small cell lung carcinoma-derived cell line
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.