missing translation for 'onlineSavingsMsg'
Learn More

Lunatic Fringe Antibody (3D12), Novus Biologicals™

Product Code. 18368037 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368037 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368037 Supplier Novus Biologicals Supplier No. H00003955M06

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lunatic Fringe Monoclonal antibody specifically detects Lunatic Fringe in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lunatic Fringe
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D12
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14851
Gene Alias beta-1,3-N-acetylglucosaminyltransferase lunatic fringe, LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, lunatic fringe (Drosophila) homolog, lunatic fringe homolog (Drosophila), O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, SCDO3EC 2.4.1.222
Host Species Mouse
Immunogen LFNG (AAH14851, 1 a.a. ~ 250 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTPGRCCLAADIQVETFIFTDGEDEALARHTGNVVITNCSAAHSRQALSCKMAVEYDRFIESGRKWFCHVDDDNYVNLRALLRLLASYPHTRDVYVGKPSLDRPIQAMERVSENKVRPVHFWFATGGAGFCISRGLALKMSPWASGGHFMNTAERIRLPDDCTIGYIVEALLGVPLIRSGLFHSHLENLQQVPTSELHEQVTLSYGMFENKRNAVHVKGPFSVEADPSRFRSIHCHLYPDTPWCPRTAIF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3955
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.