missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One LUMITRAC™ 600 μClear™ Bottom High-Binding 96-well Microplate

Product Code. 10154961
Change view
Click to view available options
Color:
White, Clear Bottom
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10154961 White, Clear Bottom Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10154961 Supplier Greiner Bio-One Supplier No. 655094
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Free of detectable DNase, RNase, human DNA and pyrogens

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Format Standard
Lid Without Lid
Detection Method Luminescence
Array 8 x 12
Autoclavable Not Autoclavable
Packaging Inner bags of 10
Binding Type High Binding
Color White, Clear Bottom
Product Type Assay Microplate
Material Polystyrene
Volume (Metric) Well 25 to 340 μL
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Sterile
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Dimensions (L x W x H) 127.76 x 85.48 x 14.40 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.